Chiller, C 900
Proveedor: KNF
KNFA301230EA 2580 EUR
KNFA301230
Chiller, C 900
This chiller is characterised by its compact design and the small footprint. The easy filling of the unit combined with splash-water proofed membrane keyboard are further benefits to simplify daily laboratory work.
Application: Rotary evaporation.
  • Low energy consumption
  • Ventilation slots at the front allow side by side placement of multiple units, saving precious lab space
  • Safe operation via splash-proof membrane keypad with LED temperature display
  • Filling level indicator for easy filling
  • External pump connections enable constant-temperature cooling of applications in an external loop
Filling volume: 1,7 to 2,6 L
Cooling agent: R134a
Pump connections: M10x1
Información de suministro: Supplied with two hose connectors for 1/4” and 3/8” tubing.