Multi-channel pipettes, electronic, variable volume, Finnpipette™ Novus i
Proveedor: Thermo Scientific
613-2276EA 2110 EUR
LABS991SP12 LABS46302100
Multi-channel pipettes, electronic, variable volume, Finnpipette™ Novus i
These electronic pipettes and the adjustable pipette stand assist in lower throughput MSIA sample enrichment. Pipettes are available in 12-channel which when combined with the adjustable pipette stand, provides an economical semi-automated system for rapid sample processing. The pipette programming is further simplified through a customised MSIA menu.
  • Semi-automated: Simplify manual sample processing using Finnpipette™ Novus i pipette on the pipette stand
  • Repetitive cycling function: New software allows 1-999 cycles with one press on the pipette trigger
  • Easy battery charging: Continuous battery charging of pipette on the adjustable stand during sample processing
  • Multiple functions and speeds: Include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more; nine aspirate/dispense speeds
  • To be used with the MSIA™ D.A.R.T.'S™ pipette tips
Información para pedidos: Pipettes are available with or without pipette stand. The tips need to be ordered separately.