Phosphatase Inhibitor Cocktail I is used to inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases.
- Storage Temperature: −30 to −10 °C
- Physical form: Liquid
- Appearance: Colourless
- MDL No.: MFCD01635520
This product thas been tested on a wide variety of cells and tissue extracts to isolate/maintain intact proteins without degradation.
Información de suministro: Contains imidazole, sodium fluoride, sodium molybdate, sodium orthovanadate, sodium pyrophosphate, and tartrate. 100X concentrate.
Precaución: This cocktail will inhibit acid-, alkaline-, and tyrosine phosphatases and is suitable for cell and tissue lysis. Store at −20 °C