Buffer, SSPE (20X)

Proveedor: Thermo Fisher Scientific
J61214.K3EA 65.7 EUR
ALFAJ61214.K3 ALFAJ61214.K2
Buffer, SSPE (20X)
Physical Form: Liquid

MDL No.: MFCD00132372
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR