Benzene-1,2-dithiocarboxamide 97%
|
Peligro
|
H51837.06EA
177
EUR
H51837.06
Benzene-1,2-dithiocarboxamide 97%
Benzene-1,2-dithiocarboxamide
Formula:
C₈H₈N₂S₂ MW: 196,28 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16293897 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...