Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide (5% BaTiO₃) in isopropanol (IPA) , 99.5% (metals basis)
Proveedor: Thermo Scientific
|
Peligro
|
43893.ACEA
168
EUR
43893.AC
Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide (5% BaTiO₃) in isopropanol (IPA) , 99.5% (metals basis)
Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide
Formula:
C₂₃H₅₀BaO₇Ti MW: 623,89 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01310249 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...