Hand disinfectants, KLEENEX®
Proveedor: Kimberly-Clark
Kleenex®
KIMB6383EA 154 EUR
KIMB6383 KIMB6382 KIMB6381
Hand disinfectants, KLEENEX®
Limpieza de la piel Desinfectantes de la piel Desinfectantes para la piel líquidos y en espuma
The alcohol-based hand sanitiser helps to reduce contamination and, without water, in hygiene-critical areas such as healthcare and food processing environment.
  • Kills (in as little as 30 seconds) up to 99,999% of the common bacteria, fungi, yeast and viruses that can cause infection or illness
  • Absorbs quickly and leaving skin feeling smooth and soft
  • Contains aloe vera and vitamins E and B
  • Colourant-free, fragrance-free
  • Pump-bottle is recyclable, when removing pump
  • With an optional bracket for convenient wall mounting
Certificaciones: Biocidal Products Directive (98/8/EC).
Precaución: Use biocides safely. Always read the label and product information before use.