Drying ovens with forced convection, TR series
Proveedor: Nabertherm
455-0546EA 7460 EUR
455-0546 455-0536 NABETR-032RN 455-0547 455-0537 455-0548 455-0538 455-0549 455-0539 455-0550 455-0551 455-0540 455-0552 455-0541 455-0542 455-0543 455-0544 455-0545 NABETR-032BN NABETR-032CN
Drying ovens with forced convection, TR series
Hornos Hornos de convección natural y con ventilación
The TR series of ovens has a maximum working temperature of up to 300 °C, horizontal forced air circulation and achieve a temperature uniformity better than ±5 °C. TR 30 to TR 240 units have been designed as bench top models, TR 450 and TR 1050 are floor standing models. Units have a large, wide-opening swing door, hinged on the right with quick release for models TR 30 - TR 450 and double swing door for TR 1050 models. A large door handle and AISI alloy 304 (DIN material no. 1.4301) stainless steel chamber, which is rust-resistant and easy to clean. They can be used for various applications such as e.g. drying, sterilising or warm storing. TR 1050 models feature transport rollers as standard, these can be ordered as an option for TR 450 models. TRS ovens for sample containing liquid solvents
  • Working temperature range: +5 °C above room temperature up to 300 °C
  • Charging in multiple layers is possible using removeable grids
  • Infinitely adjustable exhaust at the rear wall with operation from the front
  • PID microprocessor control with self-diagnosis system
  • Solid state relays ensure low noise operation
  • Temperature uniformity up to ±4 °C with closed exhaust flap in the empty workspace according to DIN 17052-1
Información de suministro: All models are supplied with a choice of controllers except TRS units which have the C450 controller. Choice of digital R 7 controller with one holding temperature, Standard B 410 controller with LC display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions or C 450 controller with LC display, 10 freely adjustable programs and 20 segments and 2 extra functions. The B 410/ C 450 controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. Note TR 450 and 1050 as well as all TRS models have 400 V 3/N/PE, other units are supplied with plugs and require a 230 V single phase power supply). A selection of accessories is available for example removeable grids with rails or reinforced trays for maximum load of 80 kg per shelf and factory fitted options such as observation window, side inlet, stainless steel collecting pan, infinitely adjustable fan speed, over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 to protect the oven and load, safety technology according to EN 1539 for charges containing liquid solvents (TRS) up to model TRS 240 which can achieve a temperature uniformity ±8 °C, optional upgrade to meet AMS 2750 E or FDA and process control and documentation with VCD software package. Please contact your Avantor Sales office for more details of these options.