Anti-HMGB3 Rabbit Polyclonal Antibody
Proveedor: BosterBio
PB9633
BSBTPB9633EA 550 EUR
BSBTPB9633
Anti-HMGB3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for High mobility group protein B3(HMGB3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
High-mobility group protein B, also known as HMG4, is a protein that in humans is encoded by the HMGB3 gene. This gene encodes a member of a family of proteins containing one or more high mobility group DNA-binding motifs. The encoded protein plays an important role in maintaining stem cell populations, and may be aberrantly expressed in tumor cells. A mutation in this gene was associated with microphthalmia, syndromic 13. There are numerous pseudogenes of this gene on multiple chromosomes. Alternative splicing results in multiple transcript variants.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.