Anti-HSPB2 Antibody

Proveedor: BosterBio
PB9598
BSBTPB9598EA 579 EUR
BSBTPB9598
Anti-HSPB2 Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Heat shock protein beta-2(HSPB2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: HSPB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR