Anti-IL6 Mouse Monoclonal Antibody Pair
Proveedor: Abnova
H00003569-AP41
ABNOH00003569-AP41EA 835 EUR
ABNOH00003569-AP41
Anti-IL6 Mouse Monoclonal Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human IL6.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-IL6, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-IL6, IgG2b kappa (50 ?g);