Anti-APOM Mouse Monoclonal Antibody Pair
Proveedor: Abnova
H00055937-AP41
ABNOH00055937-AP41EA 835 EUR
ABNOH00055937-AP41
Anti-APOM Mouse Monoclonal Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human APOM.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-APOM, IgG2b kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-APOM, IgG1 kappa (50 ?g);