Anti-SERPINA6 Polyclonal Antibody Pair
Proveedor: Abnova
H00000866-AP22
ABNOH00000866-AP22EA 835 EUR
ABNOH00000866-AP22
Anti-SERPINA6 Polyclonal Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human SERPINA6.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-SERPINA6 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-SERPINA6 (20 µg);
Type:
Antigen: SERPINA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human