Anti-Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6 Mouse Monoclonal Antibody

Proveedor: Abnova
H00009459-PW1
ABNOH00009459-PW1EA 835 EUR
ABNOH00009459-PW1
Anti-Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6 Mouse Monoclonal Antibody
Anticuerpos
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-ARHGEF6 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-ARHGEF6 (50 ?g)

Type:
Antigen: Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR