Anti-MAPK11 Polyclonal Antibody Pair

Proveedor: Abnova
H00005600-AP22
ABNOH00005600-AP22EA 835 EUR
ABNOH00005600-AP22
Anti-MAPK11 Polyclonal Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human MAPK11.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-MAPK11 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-MAPK11 (20 µg);

Type:
Antigen: MAPK11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR