Anti-PEBP1 Mouse Monoclonal Antibody (Biotin)

Proveedor: Abnova
H00005037-AP45
ABNOH00005037-AP45EA 835 EUR
ABNOH00005037-AP45
Anti-PEBP1 Mouse Monoclonal Antibody (Biotin)
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human PEBP1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PEBP1, IgG Mix kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-PEBP1, IgG1 kappa (50 ?g);

Type:
Antigen: PEBP1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR