Anti-AHSA1 Polyclonal Antibody Pair

Proveedor: Abnova
H00010598-AP22
ABNOH00010598-AP22EA 835 EUR
ABNOH00010598-AP22
Anti-AHSA1 Polyclonal Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human AHSA1.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AHSA1 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-AHSA1 (20 µg);

Type:
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR