Anti-KIT Mouse Monoclonal Antibody (Biotin)

Proveedor: Abnova
H00003815-AP43
ABNOH00003815-AP43EA 835 EUR
ABNOH00003815-AP43
Anti-KIT Mouse Monoclonal Antibody (Biotin)
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human KIT.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-KIT (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-KIT, IgG1 kappa (50 ?g);

Type:
Antigen: KIT
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR