Anti-GEMIN7 Antibody Pair

Proveedor: Abnova
H00079760-PW1
ABNOH00079760-PW1EA 835 EUR
ABNOH00079760-PW1
Anti-GEMIN7 Antibody Pair
Anticuerpos
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-GEMIN7 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-GEMIN7 (50 µl)

Type:
Antigen: GEMIN7
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR