Anti-SPP1 Mouse Monoclonal Antibody (Biotin)

Proveedor: Abnova
H00006696-AP45
ABNOH00006696-AP45EA 835 EUR
ABNOH00006696-AP45
Anti-SPP1 Mouse Monoclonal Antibody (Biotin)
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human SPP1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-SPP1 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-SPP1, IgG2b kappa (50 ?g);

Type:
Antigen: SPP1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR