Anti-TGFBI Rabbit Polyclonal Antibody

Proveedor: Abnova
H00007045-PW1
ABNOH00007045-PW1EA 835 EUR
ABNOH00007045-PW1
Anti-TGFBI Rabbit Polyclonal Antibody
Anticuerpos
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

  • Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-TGFBI (300 μl); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-TGFBI (50 μl)

Type:
Antigen: TGFBI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR