Anti-EGFR + PIK3R1 Antibody Pair

Proveedor: Abnova
ABNODI0246EA 668 EUR
ABNODI0246
Anti-EGFR + PIK3R1 Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the EGFR protein, and the other against the PIK3R1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. EGFR Rabbit purified Polyclonal Antibody (20 µg) 2. PIK3R1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: EGFR + PIK3R1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR