Anti-ESD Antibody Pair

Proveedor: Abnova
ABNOH00002098-AP51EA 835 EUR
ABNOH00002098-AP51
Anti-ESD Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human ESD.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-ESD (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-ESD (50 µg);

Type:
Antigen: ESD
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR