Anti-MRRF Antibody Pair

Proveedor: Abnova
H00092399-AP51
ABNOH00092399-AP51EA 835 EUR
ABNOH00092399-AP51
Anti-MRRF Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human MRRF.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-MRRF (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-MRRF (50 µg);

Type:
Antigen: MRRF
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR