Anti-STK11 + CAB39 Polyclonal Antibody Pair

Proveedor: Abnova
DI0569
ABNODI0569EA 668 EUR
ABNODI0569
Anti-STK11 + CAB39 Polyclonal Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the STK11 protein, and the other against the CAB39 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. STK11 Rabbit purified Polyclonal Antibody (20 µg) 2. CAB39 Mouse purified Polyclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: STK11 + CAB39
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR