Anti-SERPINA3 Antibody Pair

Proveedor: Abnova
H00000012-AP51
ABNOH00000012-AP51EA 835 EUR
ABNOH00000012-AP51
Anti-SERPINA3 Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human SERPINA3.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-SERPINA3 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-SERPINA3 (50 µg);

Type:
Antigen: SERPINA3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR