Anti-GSK3B + AXIN1 Antibody Pair

Proveedor: Abnova
DI0424
ABNODI0424EA 668 EUR
ABNODI0424
Anti-GSK3B + AXIN1 Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the GSK3B protein, and the other against the AXIN1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. GSK3B Rabbit purified Polyclonal Antibody (20 µg) 2. AXIN1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: GSK3B + AXIN1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR