Anti-PDGFA + PLCG1 Antibody Pair

Proveedor: Abnova
DI0045
ABNODI0045EA 668 EUR
ABNODI0045
Anti-PDGFA + PLCG1 Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the PDGFA protein, and the other against the PLCG1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. PDGFA Rabbit purified Polyclonal Antibody (20 µg) 2. PLCG1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PDGFA + PLCG1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR