Anti-ICOSL Antibody Pair

Proveedor: Abnova
H00023308-AP11
ABNOH00023308-AP11EA 835 EUR
ABNOH00023308-AP11
Anti-ICOSL Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human ICOSLG.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ICOSLG (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-ICOSLG, IgG1 kappa (20 µg);

Type:
Antigen: ICOSL
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR