Anti-HOXC8 Antibody Pair

Proveedor: Abnova
ABNOH00003224-AP51EA 835 EUR
ABNOH00003224-AP51
Anti-HOXC8 Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human HOXC8.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-HOXC8 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-HOXC8 (50 µg);

Type:
Antigen: HOXC8
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR