Anti-TOLLIP Antibody Pair

Proveedor: Abnova
H00054472-AP11
ABNOH00054472-AP11EA 835 EUR
ABNOH00054472-AP11
Anti-TOLLIP Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human TOLLIP.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TOLLIP (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-TOLLIP, IgG1 kappa (20 µg);

Type:
Antigen: TOLLIP
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR