Anti-OCLN + CSNK1E Antibody Pair

Proveedor: Abnova
DI0134
ABNODI0134EA 668 EUR
ABNODI0134
Anti-OCLN + CSNK1E Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the OCLN protein, and the other against the CSNK1E protein for use in Proximity Ligation Assay

Antibody pair set content: 1. OCLN Rabbit purified Polyclonal Antibody (20 µg) 2. CSNK1E Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: OCLN + CSNK1E
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR