Anti-AKT1 + SMAD4 Antibody Pair

Proveedor: Abnova
DI0011
ABNODI0011EA 668 EUR
ABNODI0011
Anti-AKT1 + SMAD4 Antibody Pair
Anticuerpos
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the AKT1 protein, and the other against the SMAD4 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. AKT1 Rabbit purified Polyclonal Antibody (20 µg) 2. SMAD4 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: AKT1 + SMAD4
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR