Anti-AKT1 Antibody Pair

Proveedor: Abnova
H00000207-AP11
ABNOH00000207-AP11EA 835 EUR
ABNOH00000207-AP11
Anti-AKT1 Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human AKT1.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AKT1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-AKT1, IgG2a kappa (20 µg);

Type:
Antigen: AKT1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR