Anti-SMAD1 + SMAD1 Antibody Pair

Proveedor: Abnova
DP0239
ABNODP0239EA 735 EUR
ABNODP0239
Anti-SMAD1 + SMAD1 Antibody Pair
Anticuerpos
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the SMAD1 protein, and the other against the specific S465 phosphorylated site of SMAD1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-SMAD1 S465 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. SMAD1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: SMAD1 + SMAD1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR