Anti-CD209 Antibody Pair

Proveedor: Abnova
H00030835-AP11
ABNOH00030835-AP11EA 835 EUR
ABNOH00030835-AP11
Anti-CD209 Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human CD209.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CD209 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-CD209, IgG2a kappa (20 µg);

Type:
Antigen: CD209
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR