Anti-KDR Antibody Pair

Proveedor: Abnova
DP0101
ABNODP0101EA 735 EUR
ABNODP0101
Anti-KDR Antibody Pair
Anticuerpos
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the KDR protein, and the other against the specific Y1175 phosphorylated site of KDR for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-KDR Y1175 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. KDR Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: KDR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR