Anti-CDKN1B Antibody Pair

Proveedor: Abnova
DP0086
ABNODP0086EA 735 EUR
ABNODP0086
Anti-CDKN1B Antibody Pair
Anticuerpos
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the CDKN1B protein, and the other against the specific T187 phosphorylated site of CDKN1B for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-CDKN1B T187 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. CDKN1B Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CDKN1B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR