Anti-RPS6KA1 Antibody Pair

Proveedor: Abnova
DP0074
ABNODP0074EA 735 EUR
ABNODP0074
Anti-RPS6KA1 Antibody Pair
Anticuerpos
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the RPS6KA1 protein, and the other against the specific T359 phosphorylated site of RPS6KA1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-RPS6KA1 T359 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. RPS6KA1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RPS6KA1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR