Anti-MAP3K5 Antibody Pair

Proveedor: Abnova
DP0038
ABNODP0038EA 735 EUR
ABNODP0038
Anti-MAP3K5 Antibody Pair
Anticuerpos
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the MAP3K5 protein, and the other against the specific S1033 phosphorylated site of MAP3K5 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-MAP3K5 S1033 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. MAP3K5 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: MAP3K5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR