Anti-CA3 Antibody Pair

Proveedor: Abnova
H00000761-AP11
ABNOH00000761-AP11EA 835 EUR
ABNOH00000761-AP11
Anti-CA3 Antibody Pair
Anticuerpos
This Set comes with matched Antibody Pair to detect and quantify protein level of human CA3.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CA3 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-CA3, IgG1 kappa (20 µg);

Type:
Antigen: CA3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR