Anti-TNF Receptor II Rabbit Polyclonal Antibody

Proveedor: BosterBio
PB9515
BSBTPB9515EA 577 EUR
BSBTPB9515
Anti-TNF Receptor II Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 1B (TNFRSF1B) detection. Tested with WB, IHC-P in Mouse;Rat.

Type: Primary
Antigen: TNF Receptor II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR