Anti-STAT5b Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9512EA 579 EUR
BSBTPB9512
Anti-STAT5b Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 5B (STAT5B) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: STAT5b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR