Anti-SFRS3 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9509EA 579 EUR
BSBTPB9509
Anti-SFRS3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Serine/arginine-rich splicing factor 3(SRSF3) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: SFRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR