Anti-SP3 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9508EA 550 EUR
BSBTPB9508
Anti-SP3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Transcription factor Sp3(SP3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: SP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR