Anti-PINX1 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9498EA 579 EUR
BSBTPB9498
Anti-PINX1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for PIN2/TERF1-interacting telomerase inhibitor 1(PINX1) detection. Tested with WB in Human.

Type: Primary
Antigen: PINX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR