Anti-NQO1 Rabbit Polyclonal Antibody

Proveedor: BosterBio
PB9497
BSBTPB9497EA 579 EUR
BSBTPB9497
Anti-NQO1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for NAD(P)H dehydrogenase [quinone] 1(NQO1) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR