Anti-ITCH/AIP4 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9496EA 579 EUR
BSBTPB9496
Anti-ITCH/AIP4 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for E3 ubiquitin-protein ligase Itchy homolog(ITCH) detection. Tested with WB in Human.

Type: Primary
Antigen: ITCH/AIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR