Anti-CPT1B Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9491EA 550 EUR
BSBTPB9491
Anti-CPT1B Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Carnitine O-palmitoyltransferase 1, muscle isoform (CPT1B) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR