Anti-CIB1 Rabbit Polyclonal Antibody

Proveedor: BosterBio
PB9489
BSBTPB9489EA 579 EUR
BSBTPB9489
Anti-CIB1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Calcium and integrin-binding protein 1(CIB1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: CIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR